Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichloro-4-fluorophenol
2,6-Dichloro-4-fluorophenol
2,6-Dichloro-4-fluorophenol
2,6-Dichloro-4-fluorophenol
Supplier:
FLUOROCHEM
Certificates
Expand image
2,6-Dichloro-4-fluorophenol
Supplier:
FLUOROCHEM
About this item
CAS Number:
392-71-2
:
Formula:
C₆H₃Cl₂FO
MDL Number:
MFCD00010675
:
180.99 g/mol
Boiling Pt:
142 °C (100 mmHg)
Melting Pt:
50 °C
UN:
3261
ADR:
8,III
:
GHS07