
3-Chloro-4-fluoroaniline for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:367-21-5
:
Formula:C₆H₅ClFN
MDL Number:MFCD00007767
:145.56 g/mol
Boiling Pt:230 °C (1013 hPa)
Melting Pt:41…44 °C
Density:1.226 g/cm³ (20 °C)
EINECS:206-682-8
ADR:6.1,III
:M-Clarity™ quality level = MQ200
UN:2811
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:10 g/l (20 °C)
- Melting Point:41 - 44 °C
- Molar Mass:145.56 g/mol
- Bulk Density:1000 kg/m3
- Boiling Point:230 °C (1013 hPa)
- Vapor Pressure:0.01 hPa (20 °C)
- Flash Point:114 °C
- Density:1.59 g/cm3
- pH:7 (10 g/l, H2O, 20 °C)