Order Entry
Germany
ContactUsLinkComponent
2-Chloro-6-fluoroaniline 99%
2-Chloro-6-fluoroaniline 99%
Certificates
undefined
2-Chloro-6-fluoroaniline 99%

Specification Test Results

  • Appearance:
    Clear light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5480 to 1.5540 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information