


2-Chloro-4-fluoroaniline ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2106-02-7
:
Formula:C₆H₅ClFN
MDL Number:MFCD00042530
:145.56 g/mol
Boiling Pt:191…192 °C
Density:1.219
Flash Pt:98 °C (208 °F)
EINECS:218-282-0
UN:2810
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient