Chlorobenzene
Catalog # KRAE30-160.5000
Supplier: KRAEMER MARTIN
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:112.56 g/mol
Merck Index:13,02139
Boiling Pt:130 °C (1013 hPa)
Melting Pt:–45 °C
Density:1.11 g/cm³ (20 °C)
Flash Pt:28 °C
Storage Temperature:Ambient
EINECS:203-628-5
UN:1134
ADR:3,III
REACH:01-2119432722-45
:GHS02
:GHS07
:GHS09
CAS Number:108-90-7
Formula:C₆H₅Cl
MDL Number:MFCD00000530