Order Entry
Germany
ContactUsLinkComponent
Photographic trays, LaboPlast®
Photographic trays, LaboPlast®
Catalog # BURK4206-2030
Supplier:  BURKLE
undefined
Photographic trays, LaboPlast®
Catalog # BURK4206-2030
Supplier:  BURKLE

Specifications

  • Colour:
    Red
  • Height:
    105 mm
  • Base:
    310×410 mm
  • Pk:
    1

Specifications

About this item

Made of PVC.

  • Thick sides, shatterproof
  • Largely resistant to acids and alkalis
  • Not suitable for temperature controlled cupboards or dishwashers
  • Temperature resistant from −30 °C up to +60 °C max.