Orders LinkContactUsLinkComponent
Venipuncture sets, Surflo®
Venipuncture sets, Surflo®
Supplier:   TERUMO
Venipuncture sets, Surflo®
Supplier:  TERUMO
Certificates
Venipuncture sets, Surflo®
3 Options Available Below
Product Details & Documents
Surshield prevents needle stick injuries with painless placement.
Cannula sharpness combined with a special silicone coating allows an easy and smooth placement.
Product Family Options
Product Information
  • Gauge
    Colour
    Length
    Availability
    Price
  • TERUSV-S19NL30
    Winged needles with protector
    Winged needles with protector
    G 19
    White
    19 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 55,40 €
    55,40 €
    1 * 50 ST
       55,40 €
  • TERUSV-S21NL30
    Winged needles with protector
    Winged needles with protector
    G 21
    Green
    19 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 55,40 €
    55,40 €
    1 * 50 ST
       55,40 €
  • TERUSV-S23NL30
    Winged infusion sets with protector
    Winged infusion sets with protector
    G 23
    Light blue
    19 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 55,40 €
    55,40 €
    1 * 50 ST
       55,40 €