p-Anisic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-09-4
:
Formula:C₈H₈O₃
MDL Number:MFCD00002542
:152.15 g/mol
Merck Index:13,00669
Boiling Pt:275 °C (760 mmHg)
Melting Pt:181…186 °C
Density:~ 1,385 g/cm³ (20 °C)
Flash Pt:185 °C
Storage Temperature:Ambient
EINECS:202-818-5
Specification Test Results
- Appearance:White to beige to light grey Powder
- Infrared spectrum:Conforms
- Melting point:181 to 186 °C
- GC:≥97.5 %