Order Entry
Germany
ContactUsLinkComponent
3-Cyanobenzoic acid 98%
Irritant
3-Cyanobenzoic acid 98%
Certificates
undefined
Irritant
3-Cyanobenzoic acid 98%

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    217 to 222 °C
  • HPLC:
    ≥97.5 %

Specification Test Results

Related Information