DSC calibration standard, benzoic acid
Catalog # PROOLGC2606
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Description:Benzoic acid
- Seq:01
- Melting pt.:122.35 °C
Specifications
About this item
Enthalpy of fusion: 17.98 kJ/mol