0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Benzoic acid methyl ester
Benzoic acid methyl ester ≥99%
Benzoic acid methyl ester ≥99%
:
Thermo Fisher Scientific
Benzoic acid methyl ester ≥99%
:
Thermo Fisher Scientific
:
93-58-3
:
:
C₆H₅COOCH₃
:
MFCD00008421
:
136.15 g/mol
:
14,6024
:
198…200 °C
:
–12 °C
:
1.094
:
82 °C (179 °F)
:
202-259-7
:
GHS07
:
Ambient
msds_ALFAA12404
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years