4-Aminobenzonitrile
MOLE30105118-25G
: Molekula
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
:
:118.14 g/mol
:167 °C (1.1 mmHg)
:84…87 °C
:1.274 g/cm³ (20 °C)
:Ambient
:212-850-1
:3439
:6.1,III
:GHS07
:873-74-5
:C₇H₆N₂
:MFCD00007821