4-Aminobenzonitrile
Supplier: CHEMPUR
Certificates
About this item
CAS Number:873-74-5
:
Formula:C₇H₆N₂
MDL Number:MFCD00007821
:118.14 g/mol
Boiling Pt:167 °C (1.1 mmHg)
Melting Pt:84…87 °C
Density:1.274 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-850-1
UN:3439
ADR:6.1,III
:GHS07