Order Entry
Germany
ContactUsLinkComponent
3-Aminobenzonitrile
3-Aminobenzonitrile
Catalog # MOLE62393991-25G
Supplier:  Molekula
CAS Number:  
3-Aminobenzonitrile
Catalog # MOLE62393991-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Boiling Pt:288…290 °C (1013 hPa)
Melting Pt:48…50 °C
Density:1.14 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:6.1,III
 :118.14 g/mol
EINECS:218-800-5
UN:3439
CAS Number:2237-30-1
Formula:C₇H₆N₂
MDL Number:MFCD00007756