
2,5-Dimethoxybenzaldehyde 97%
:93-02-7
:
:C₉H₁₀O₃
:MFCD00003314
:166.18 g/mol
:146 °C (10 mmHg)
:45…50 °C
:113 °C
:Ambient
:202-211-5
:GHS07
- Appearance:Yellow to beige Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:46 to 51 °C
- Water:≤0.5 %
- GC:≥96.0 %