3-Hydroxy-p-anisaldehyde, Sigma-Aldrich®
Catalog # SIAL59940-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:152.15 g/mol
Boiling Pt:308 °C (1013 hPa)
Melting Pt:68…70 °C
Density:1.401 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-694-9
CAS Number:621-59-0
Formula:C₈H₈O₃
MDL Number:MFCD00003369