Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Methoxysalicylaldehyde
5-Methoxysalicylaldehyde
5-Methoxysalicylaldehyde
MOLE22481955-5G
:
Molekula
:
5-Methoxysalicylaldehyde
MOLE22481955-5G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
248 °C (1013 hPa)
:
51…52 °C
:
1.21 g/cm³ (20 °C)
:
152.15 g/mol
:
Ambient
:
211-589-0
:
672-13-9
:
C₈H₈O₃
:
MFCD00003332