Blotting papers
Their extremely smooth surface and relatively high thickness provide them with superior absorption properties and uniformity across the entire contact area.
- Grades BF 2 and BF 3 are made of cotton linters with more than 98% alpha-cellulose content
- Four grades of blotting papers are available
Grades BF 1 and BF 4 are made of refined pulp and cotton linters with more than 95% alpha-cellulose content.
Their extremely smooth surface and relatively high thickness provide them with superior absorption properties and uniformity across the entire contact area.
- Grades BF 2 and BF 3 are made of cotton linters with more than 98% alpha-cellulose content
- Four grades of blotting papers are available
Grades BF 1 and BF 4 are made of refined pulp and cotton linters with more than 95% alpha-cellulose content.
- Catalog No:
- SARTFT-2519200200N
- 588-3851
- 588-3852
- SARTFT2-520200200N
- SARTFT2520-460570K
- 515-1606
- 515-1632
- SARTFT-3-521-185
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Weight:
- 195 g/m²
- 195 g/m²
- 195 g/m²
- 330 g/m²
- 330 g/m²
- 330 g/m²
- 550 g/m²
- 550 g/m²
- Thickness:
- 0,35 mm
- 0,35 mm
- 0,35 mm
- 0,76 mm
- 0,76 mm
- 0,76 mm
- 1,3 mm
- 1,3 mm
- Environmental certification:
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
- Renewable Energy Certificate
Product Family Options
Grade BF 2
- DescriptionDimensions
Low Manufacturing Impact Specifications
Cat. No.SARTFT-2519200200NEnvironmentally PreferableLow Manufacturing ImpactWeight195 g/m²Thickness0,35 mmEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.588-3851Environmentally PreferableLow Manufacturing ImpactWeight195 g/m²Thickness0,35 mmEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.588-3852Environmentally PreferableLow Manufacturing ImpactWeight195 g/m²Thickness0,35 mmEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact
Grade BF 3
- DescriptionDimensions
Low Manufacturing Impact Specifications
Cat. No.SARTFT2-520200200NEnvironmentally PreferableLow Manufacturing ImpactWeight330 g/m²Thickness0,76 mmEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.SARTFT2520-460570KEnvironmentally PreferableLow Manufacturing ImpactWeight330 g/m²Thickness0,76 mmEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.515-1606Environmentally PreferableLow Manufacturing ImpactWeight330 g/m²Thickness0,76 mmEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact
Grade BF 4
- DescriptionDimensions
Low Manufacturing Impact Specifications
Cat. No.515-1632Environmentally PreferableLow Manufacturing ImpactWeight550 g/m²Thickness1,3 mmEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.SARTFT-3-521-185Environmentally PreferableLow Manufacturing ImpactWeight550 g/m²Thickness1,3 mmEnvironmental certificationRenewable Energy CertificateLow Manufacturing Impact