Order Entry
Germany
Orders LinkContactUsLinkComponent
3-Nitrobenzaldehyde

3-Nitrobenzaldehyde

Glass bottle for solids, 500 g
Catalog # MOLEM63370135
Marketsource
Supplier:   Molekula
3-Nitrobenzaldehyde
Catalog # MOLEM63370135
Supplier:  Molekula
Marketsource
CAS Number:  99-61-6
3-Nitrobenzaldehyde

Glass bottle for solids, 500 g

CAS Number:  99-61-6
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:99-61-6
  • Formula:C₇H₅NO₃
  • MDL Number:MFCD00007249
  • Molecular Weight:151.12 g/mol
  • Merck Index:12,06684
  • Boiling Pt:232 °C (1013 hPa)
  • Melting Pt:58 °C
  • Density:1.481 g/cm³ (0 °C)
  • Storage Temperature:Ambient
  • EINECS:202-772-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications