3-Nitrobenzaldehyde
Marketsource
Supplier: CHEMPUR
Jump to:
About this item
- CAS Number:99-61-6
- Formula:C₇H₅NO₃
- MDL Number:MFCD00007249
- Molecular Weight:151.12 g/mol
- Merck Index:12,06684
- Boiling Pt:232 °C (1013 hPa)
- Melting Pt:58 °C
- Density:1.481 g/cm³ (0 °C)
- Storage Temperature:Ambient
- EINECS:202-772-6
2 Options Available Below
Product Details & Documents
- CAS Number:99-61-6
- Formula:C₇H₅NO₃
- MDL Number:MFCD00007249
- Molecular Weight:151.12 g/mol
- Merck Index:12,06684
- Boiling Pt:232 °C (1013 hPa)
- Melting Pt:58 °C
- Density:1.481 g/cm³ (0 °C)
- Storage Temperature:Ambient
- EINECS:202-772-6
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource



