Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bottles
Plastic Bottles
Bottles, wide neck, PFA, with screw cap
Bottles, wide neck, PFA, with screw cap
Bottles, wide neck, PFA, with screw cap
Supplier:
KARTELL
Certificates
Expand image
Bottles, wide neck, PFA, with screw cap
Supplier:
KARTELL
Certificates
About this item
PFA, translucent.
Temperature resistance: from −200 to +250 °C
Universal chemical resistance
Pressure: no pressure
Non porous
Related Information
Documentation
Certificates
Search by Lot Number
Documentation