Bottles, narrow neck, round, HDPE, with screw caps
Supplier: KARTELL
Certificates
About this item
HDPE, translucent.
- Good chemical resistance
- Good temperature resistance
- Suitable for foodstuff
The cylindrical bottles are supplied with insert and cap. Thanks to the large neck they are suitable for powders, solid specimen, creamy or semisolid substances, crystals and hygroscopic substances.
Certifications: EC Regulation 1935/2004 • EC Regulation 10/2011