Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dibromo-5-fluoro-2-iodobenzene
1,3-Dibromo-5-fluoro-2-iodobenzene
1,3-Dibromo-5-fluoro-2-iodobenzene
1,3-Dibromo-5-fluoro-2-iodobenzene
Supplier:
FLUOROCHEM
Certificates
Expand image
1,3-Dibromo-5-fluoro-2-iodobenzene
Supplier:
FLUOROCHEM
About this item
CAS Number:
62720-29-0
:
Formula:
C₆H₂Br₂FI
MDL Number:
MFCD00042232
Boiling Pt:
286 °C (1013 hPa)
Melting Pt:
135…135,6 °C
:
379.79 g/mol
:
GHS07