Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl (trimethylsilyl)acetate
Ethyl (trimethylsilyl)acetate
Ethyl (trimethylsilyl)acetate
Ethyl (trimethylsilyl)acetate
Supplier:
CHEMPUR SILANE
Certificates
Expand image
Ethyl (trimethylsilyl)acetate
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
4071-88-9
:
Formula:
C₇H₁₆O₂Si
MDL Number:
MFCD00009172
ADR:
3,III
:
160.29 g/mol
EINECS:
223-783-2
UN:
3272