Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Methoxy-1-propanol
3-Methoxy-1-propanol, Sigma-Aldrich®
3-Methoxy-1-propanol, Sigma-Aldrich®
SIAL38457-10ML-F
:
Merck
:
3-Methoxy-1-propanol, Sigma-Aldrich®
SIAL38457-10ML-F
:
Merck
:
Pack type:
Glass bottle
Pk:
10 mL
Seq:
2275
:
:
90.12 g/mol
:
433-230-9
:
1987
:
3,III
:
1589-49-7
:
C₄H₁₀O₂
:
MFCD00039550