Order Entry
Germany
Orders LinkContactUsLinkComponent
D-Cycloserine

D-Cycloserine

Glass bottle for solids, 1 g
Catalog # SERA17685.01
Irritant
Marketsource
Supplier:   Serva
D-Cycloserine
Catalog # SERA17685.01
Supplier:  Serva
Marketsource
CAS Number:  68-41-7
D-Cycloserine

Glass bottle for solids, 1 g

CAS Number:  68-41-7
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine
  • CAS Number:68-41-7
  • Formula:C₃H₆N₂O₂
  • MDL Number:MFCD00005353
  • Molecular Weight:102.09 g/mol
  • Storage Temperature:Freezer
  • EINECS:200-688-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications