Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4,6-Tris(2-pyridyl)-1,3,5-triazine
2,4,6-Tris(2-pyridyl)-1,3,5-triazine
2,4,6-Tris(2-pyridyl)-1,3,5-triazine
2,4,6-Tris(2-pyridyl)-1,3,5-triazine
MOLEM44417371
:
Molekula
:
2,4,6-Tris(2-pyridyl)-1,3,5-triazine
MOLEM44417371
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
312.33 g/mol
:
Ambient
:
222-965-9
:
3682-35-7
:
C₁₈H₁₂N₆
:
MFCD00006045