Benzophenone hydrazone
Catalog # MOLE19039648-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:196.25 g/mol
EINECS:226-321-8
UN:2811
ADR:6.1,III
CAS Number:5350-57-2
Formula:C₁₃H₁₂N₂
MDL Number:MFCD00007624