Order Entry
Germany
Orders LinkContactUsLinkComponent
 

D-Cycloserine

Irritant
Marketsource
Supplier:   Molekula
D-Cycloserine
Supplier:  Molekula
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine
  • CAS Number:68-41-7
  • Formula:C₃H₆N₂O₂
  • MDL Number:MFCD00005353
  • Molecular Weight:102.09 g/mol
  • Storage Temperature:Freezer
  • EINECS:200-688-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

    Specifications:
    Marketsource
    Marketsource