Order Entry
Germany
ContactUsLinkComponent
Silver trifluoroacetate, Sigma-Aldrich®
Irritant
Environment
Silver trifluoroacetate, Sigma-Aldrich®
Catalog # 8.18003.0025
Supplier:  Merck
CAS Number:  
Irritant
Environment
Silver trifluoroacetate, Sigma-Aldrich®
Catalog # 8.18003.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    257 - 260 °C
  • Molar Mass:
    220.88 g/mol

Specification Test Results

About this item

 :
 :220.88 g/mol
ADR:9,III
 :M-Clarity™ quality level = MQ200
EINECS:221-004-0
UN:3077
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:2966-50-9
Formula:C₂AgF₃O₂
MDL Number:MFCD00013199