2-Amino-2-methylpropanol hydrochloride, Sigma-Aldrich®
Catalog # 8.14099.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:203 - 206 °C
- Molar Mass:125.60 g/mol
- Synonyms:AMP hydrochloride
- Hill Formula:C4H12ClNO
- Chemical Formula:(CH3)2C(NH2)CH2OH * HCl
Specification Test Results
About this item
:
:125.6 g/mol
:M-Clarity™ quality level = MQ200
EINECS:221-713-5
CAS Number:3207-12-3
Formula:C₄H₁₂ClNO
MDL Number:MFCD00039086