Order Entry
Germany
ContactUsLinkComponent
Methylammonium chloride, Sigma-Aldrich®
Irritant
Methylammonium chloride, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Methylammonium chloride, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    228 - 233 °C
  • Molar Mass:
    67.52 g/mol
  • Bulk Density:
    1100 kg/m3
  • Boiling Point:
    225 - 230 °C (20 hPa)
  • pH:
    5 - 7 (10 g/l, H2O, 20 °C)
  • Hill Formula:
    CH6ClN
  • Chemical Formula:
    CH3NH3Cl

Specification Test Results

Related Information