Order Entry
Germany
ContactUsLinkComponent
Methyl acrylate stabilised for synthesis, Sigma-Aldrich®
Flammable
Toxic
Methyl acrylate stabilised for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Toxic
Methyl acrylate stabilised for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    52 g/l (25 °C)
  • Melting Point:
    -74 °C
  • Molar Mass:
    86.09 g/mol
  • Boiling Point:
    79 - 81 °C (1013 hPa)
  • Vapor Pressure:
    93 hPa (20 °C)
  • Flash Point:
    -2.7 °C
  • Refractive Index:
    1.4021 (20 °C, 589 nm)
  • Explosion Limit:
    2.8 - 18.6 %(V)
  • Density:
    0.96 g/cm3 (20 °C)
  • Ignition Point:
    415 °C
  • Water absorption:
    250 g/kg (20 °C)
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

Related Information