Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chlorophenylsulphonylacetone
4-Chlorophenylsulphonylacetone
4-Chlorophenylsulphonylacetone
4-Chlorophenylsulphonylacetone
Supplier:
MENAI
Certificates
Expand image
4-Chlorophenylsulphonylacetone
Supplier:
MENAI
About this item
CAS Number:
5000-48-6
:
Formula:
C₉H₉ClO₃S
MDL Number:
MFCD00018663
:
232.69 g/mol