Order Entry
Germany
ContactUsLinkComponent
Organic reference standard, Dimethylnitrosoamine 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, Dimethylnitrosoamine 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA15604000ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Dimethylnitrosoamine 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA15604000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    62-75-9
  • Description:
    Dimethylnitrosoamine 100 µg/ml in methanol
  • Formula:
    C₂H₆N₂O
  • Description:
    Dimethylnitrosoamine

Specifications

About this item