Order Entry
Germany
ContactUsLinkComponent
Organic reference standard, 1,5-Dinitronaphthalene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Organic reference standard, 1,5-Dinitronaphthalene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20974500CY
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 1,5-Dinitronaphthalene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20974500CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    605-71-0
  • Description:
    1,5-Dinitronaphthalene 10 µg/ml in cyclohexane
  • Formula:
    C₁₀H₆N₂O₄
  • Description:
    1,5-Dinitronaphthalene

Specifications

About this item