Pesticide reference standard, 1,3-Dichlorobenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
EHERL12371000CY
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:541-73-1
- Description:1,3-Dichlorobenzene 10 µg/ml in cyclohexane
- Formula:C₆H₄Cl₂
- Description:1,3-Dichlorobenzene