Order Entry
Germany
ContactUsLinkComponent
Organic reference standard, Caffeine 10 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, Caffeine 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL11693000ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Caffeine 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL11693000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    58-08-2
  • Description:
    Caffeine 10 µg/ml in methanol
  • Formula:
    C₈H₁₀N₄O₂
  • Description:
    Caffeine

Specifications

About this item