Methyl pelargonate
Catalog # EHERC15623150
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:172.27 g/mol
EINECS:217-052-7
CAS Number:1731-84-6
Formula:C₁₀H₂₀O₂
MDL Number:MFCD00009569