
(±)-1-Butoxy-2-propanol
Catalog # EHERC10900100
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:132.2 g/mol
EINECS:225-878-4
:GHS07
Storage Temperature:Ambient
CAS Number:5131-66-8
Formula:C₇H₁₆O₂
MDL Number:MFCD00021904