D-Cycloserine
Marketsource
Supplier: BACHEM BIOCHEMICA
Jump to:
About this item
Synonyms: D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine
- CAS Number:68-41-7
- Formula:C₃H₆N₂O₂
- MDL Number:MFCD00005353
- Molecular Weight:102.09 g/mol
- Storage Temperature:Freezer
- EINECS:200-688-4
2 Options Available Below
Product Details & Documents
Synonyms: D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine
- CAS Number:68-41-7
- Formula:C₃H₆N₂O₂
- MDL Number:MFCD00005353
- Molecular Weight:102.09 g/mol
- Storage Temperature:Freezer
- EINECS:200-688-4
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



