Order Entry
Germany
Orders LinkContactUsLinkComponent
 

D-Cycloserine

Irritant
Supplier:   PanReac AppliChem
D-Cycloserine
Supplier:  PanReac AppliChem
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine
  • CAS Number:68-41-7
  • Formula:C₃H₆N₂O₂
  • MDL Number:MFCD00005353
  • Molecular Weight:102.09 g/mol
  • Storage Temperature:Freezer
  • EINECS:200-688-4

 

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 79,85 €
    79,85 €
    1 * 1 g
       79,85 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 284,37 €
    284,37 €
    1 * 5 g
       284,37 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 1 033,61 €
    1 033,61 €
    1 * 25 g
       1 033,61 €