4-Nitrodiphenylamine 98+%, rods
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 836-30-6 Molecular Formula: C12H10N2O2 Formula Weight: 214.22 Flash Point: 190°(374°F) Melting Point: 132-135° Boiling Point: 211°/30mm Beilstein Reference: 2051910 UN No.: 3077 MDL No.: MFCD00007301
Synonyms: 4-Nitro-N-phenylaniline
- CAS Number:836-30-6
- Formula:C₁₂H₁₀N₂O₂
- MDL Number:MFCD00007301
- Molecular Weight:214.22 g/mol
- Boiling Pt:211 °C (30 mmHg)
- Melting Pt:132…135 °C
- Flash Pt:190 °C (374 °F)
- Storage Temperature:Ambient
- EINECS:212-646-2
- UN:3077
- ADR:9,III
2 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 836-30-6 Molecular Formula: C12H10N2O2 Formula Weight: 214.22 Flash Point: 190°(374°F) Melting Point: 132-135° Boiling Point: 211°/30mm Beilstein Reference: 2051910 UN No.: 3077 MDL No.: MFCD00007301
Synonyms: 4-Nitro-N-phenylaniline
- CAS Number:836-30-6
- Formula:C₁₂H₁₀N₂O₂
- MDL Number:MFCD00007301
- Molecular Weight:214.22 g/mol
- Boiling Pt:211 °C (30 mmHg)
- Melting Pt:132…135 °C
- Flash Pt:190 °C (374 °F)
- Storage Temperature:Ambient
- EINECS:212-646-2
- UN:3077
- ADR:9,III



