Order Entry
Germany
ContactUsLinkComponent
Irritant
5-Hydroxy-2-(trifluoromethyl)pyridine 97%
Catalog # H31734.03
CAS Number:  
Irritant
5-Hydroxy-2-(trifluoromethyl)pyridine 97%
Catalog # H31734.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :163.1 g/mol
Melting Pt:174…176 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:216766-12-0
Formula:C₆H₄F₃NO
MDL Number:MFCD06801325