Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dithiooxamide
Dithiooxamide ≥98%
Dithiooxamide ≥98%
Dithiooxamide ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Dithiooxamide ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
79-40-3
:
Formula:
C₂H₄N₂S₂
MDL Number:
MFCD00004941
:
120.2 g/mol
Melting Pt:
245 °C (dec.)
EINECS:
201-203-9
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB24866
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation