
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:270573-71-2
:MFCD00007473
Formula:C₆H₄Na₂O₈S₂·1H₂O
MDL Number:MFCD00149531
:332.22 g/mol
Merck Index:14,9465
Melting Pt:>300 °C
EINECS:205-741-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together