3-Amino-3-phenyl-1-propanol 94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14593-04-5
:
Formula:C₉H₁₃NO
MDL Number:MFCD00801124
:151.21 g/mol
Melting Pt:70…77 °C
Storage Temperature:Ambient
UN:3259
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White Needles
- Infrared spectrum:Conforms
- Melting point:70 to 77 °C
- Titration with HClO4:≥92 %