Order Entry
Germany
ContactUsLinkComponent
 
 
 
2,6-Toluenediisocyanate 97%
 
undefined
 
 
2,6-Toluenediisocyanate 97%

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5700 to 1.5720 (20°C, 589 nm)
  • GC:
    ≥96.0 %

 

 

Frequently Bought Together