0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
(±)-1-Butoxy-2-propanol
(±)-1-Butoxy-2-propanol
(±)-1-Butoxy-2-propanol
Irritant
(±)-1-Butoxy-2-propanol
Supplier:
abcr
Certificates
Expand image
Irritant
(±)-1-Butoxy-2-propanol
Supplier:
abcr
About this item
CAS Number:
5131-66-8
:
Formula:
C₇H₁₆O₂
MDL Number:
MFCD00021904
:
132.2 g/mol
EINECS:
225-878-4
:
GHS07
Storage Temperature:
Ambient