Drum pumps B2 Vario PP
Supplier: LUTZ PUMPEN
Certificates
About this item
Drum pumps for thin bodied fluids such as battery acids, amonia solutions, photographic developer/-fixer, glycol, phosphoric acids, hydrochloric acid and hydorgen peroxides.For filling small quantities from hobbocks, canisters and drums
- Due to the modular construction, the pump is service-friendly
- The pumps offer a complete liquid drainage function
- Already pre-assembled: immediately ready for action
- Ideal for pumping and filling thin bodied liquids
Ordering information: Available as a set with accessories or pump-only (no hose or other accessories).
Delivery: Sets include pump, 1,5 m PVC hose, Lutz nozzle, 2 hose clips, and wall bracket.